missing translation for 'onlineSavingsMsg'
Learn More

N-Gamma-Acetyl-N-2-formyl-5-methoxykynurenamine, TRC

Product Code. 18080050
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18080050 10 mg 10mg
18043120 1 mg 1mg
18011730 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18080050 Supplier Toronto Research Chemicals Supplier No. TRCA17610010MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52450-38-1
Molecular Formula C13 H16 N2 O4
Molecular Weight (g/mol) 264.28
Synonym N-[3-[2-(Formylamino)-5-methoxyphenyl]-3-oxopropyl]acetamide, N1-Acetyl-N2-formyl-5-methoxykynuramine, NSC 688263
IUPAC Name N-[3-(2-formamido-5-methoxyphenyl)-3-oxopropyl]acetamide
SMILES COc1ccc(NC=O)c(c1)C(=O)CCNC(=O)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H16N2O4/c1-9(17)14-6-5-13(18)11-7-10(19-2)3-4-12(11)15-8-16/h3-4,7-8H,5-6H2,1-2H3,(H,14,17)(H,15,16)
Formula Weight 264.111
Percent Purity >95%
Chemical Name or Material N-gamma-Acetyl-N-2-formyl-5-methoxykynurenamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.