missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3,3-diphenylpropylamine, TRC

Product Code. 30007558
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007558 5 g 5g
30006801 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30007558 Supplier Toronto Research Chemicals Supplier No. TRCM3037105G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28075-29-8
Molecular Formula C16H19N
Molecular Weight (g/mol) 225.33
Synonym N-Methyl-gamma-phenyl-benzenepropanamine, (3,3-Diphenylpropyl)methylamine
IUPAC Name N-methyl-3,3-diphenylpropan-1-amine
SMILES CNCCC(c1ccccc1)c2ccccc2

Specifications

Quantity 5 g
InChI Formula InChI=1S/C16H19N/c1-17-13-12-16(14-8-4-2-5-9-14)15-10-6-3-7-11-15/h2-11,16-17H,12-13H2,1H3
Formula Weight 225.1517
Chemical Name or Material N-Methyl-3,3-diphenylpropylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.