missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis(4-methoxybenzyl)amine, TRC

Product Code. 30470344
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30470344 1 g 1g
30464344 5 g 5g
30472828 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30470344 Supplier Toronto Research Chemicals Supplier No. TRCM2656901G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17061-62-0
Molecular Formula C16H19NO2
Molecular Weight (g/mol) 257.33
Synonym N-(4-Methoxybenzyl)-1-(4-methoxyphenyl)methanamine, N,N-Bis(4-Methoxybenzyl)amine, Bis(4-methoxybenzyl)amine, 4,4'-dimethoxy-dibenzylamine
IUPAC Name 1-(4-methoxyphenyl)-N-[(4-methoxyphenyl)methyl]methanamine
SMILES COc1ccc(CNCc2ccc(OC)cc2)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C16H19NO2/c1-18-15-7-3-13(4-8-15)11-17-12-14-5-9-16(19-2)10-6-14/h3-10,17H,11-12H2,1-2H3
Formula Weight 257.1416
Chemical Name or Material N,N-Bis(4-methoxybenzyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.