missing translation for 'onlineSavingsMsg'
Learn More

N-Nitroso-N-methyl-4-aminobutyric Acid (200 μg/mL in Methanol), TRC

Product Code. 30003320
Change view
Click to view available options
Quantity:
1 x 1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003320 1 x 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003320 Supplier Toronto Research Chemicals Supplier No. TRCKIT84401X1ML
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 61445-55-4
Molecular Formula C5 H10 N2 O3
Quantity 1 x 1 mL
Synonym Butanoic acid, 4-(methylnitrosoamino)-, N-Nitroso-N-methyl-4-aminobutyric acid, NMBA, N-Nitrosomethylaminobutyric acid, 4-[Methyl(nitroso)amino]butanoic acid
InChI Formula InChI=1S/C5H10N2O3/c1-7(6-10)4-2-3-5(8)9/h2-4H2,1H3,(H,8,9)
SMILES CN(CCCC(=O)O)N=O
IUPAC Name 4-[methyl(nitroso)amino]butanoic acid
Molecular Weight (g/mol) 146.14
Formula Weight 146.0691
Concentration or Composition (by Analyte or Components) N-Nitroso-N-methyl-4-aminobutyric Acid 200 μg/mL
Chemical Name or Material N-Nitroso-N-methyl-4-aminobutyric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.