Learn More
N-Nitrosodicyclohexylamine, TRC
Product Code. 17915095
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17915095 50 mg 50mg
17913605 100 mg 100mg
17933295 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17915095 Supplier Toronto Research Chemicals Supplier No. TRCN52555050MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 947-92-2
Molecular Formula C12 H22 N2 O
Molecular Weight (g/mol) 210.32
Synonym N-Nitrosodicyclohexylamine, Dicyclohexylamine, N-nitroso- (6CI,7CI,8CI), N-Cyclohexyl-N-nitrosocyclohexanamine, Dicyclohexylnitrosamine, N,N-Dicyclohexylnitrosamine, NSC 337
IUPAC Name N,N-dicyclohexylnitrous amide
SMILES O=NN(C1CCCCC1)C2CCCCC2
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C12H22N2O/c15-13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h11-12H,1-10H2
Formula Weight 210.1732
Percent Purity >95%
Chemical Name or Material N-Nitrosodicyclohexylamine
Grade HPLC