missing translation for 'onlineSavingsMsg'
Learn More

N1,N12-Diethylspermine tetrahydrochloride, Tocris Bioscience™

Product Code. 18074912
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18074912 10 mg 10mg
18025242 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18074912 Supplier Tocris Bioscience Supplier No. 0500/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Polyamine synthase inhibitor

Powerful antineoplastic agent in cultured cells and animal tumors (IC50 = 0.2 μM in L1210 cells). Inhibitor of polyamine synthases. Suppressor of mitochondrial DNA synthesis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113812-15-0
Molecular Formula C14H38Cl4N4
Molecular Weight (g/mol) 404.286
InChI Key SCYOBNRPMHJGLB-UHFFFAOYSA-N
Synonym n1, n12-diethylsperminea.4hcl, n1,n12-diethylspermine tetrahydrochloride, n1, n12-diethylspermine.4hcl, n1,n12-diethylspermine tetra hydrochloride, 3,7,12,16-tetraazaoctadecane tetrahydrochloride, n1, n12-diethylspermine? inverted exclamation mark currency4hcl
PubChem CID 14056749
IUPAC Name N,N'-bis[3-(ethylamino)propyl]butane-1,4-diamine;tetrahydrochloride
SMILES CCNCCCNCCCCNCCCNCC.Cl.Cl.Cl.Cl

Specifications

Quantity 10 mg
Formula Weight 404.29
Percent Purity >98%
Chemical Name or Material N1, N12-Diethylspermine tetrahydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.