missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ N6-Methyladenosine (m6A) Monoclonal Antibody (A8F2)

Description
Product MA5-50598 was used in a Competitive ELISA.
N6-methyladenosine (m6A), or methylation of the N6 position of adenosine is a posttranscriptional modification of RNA. Due to a lack of analytical methods, N6-methyladenosine is poorly understood, but new evidence suggests that it is a very common base modification and important physiological regulator. N6-methyladenosine is markedly increased throughout brain development, and is enriched near stop codons, microRNA-binding sites and UTRs, which indicates a fundamental role in the regulation of gene expression. N6-methyladenosine is also highly conserved between human and mouse.
Specifications
Specifications
| Antigen | N6-Methyladenosine (m6A) |
| Applications | Dot Blot, ELISA |
| Classification | Monoclonal |
| Clone | A8F2 |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene Alias | C11H15N5O4; m6A |
| Host Species | Mouse |
| Immunogen | m6A Small Molecule conjugated to OVA. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?