missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NAB1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15789690 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15789690 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15789690 Supplier Invitrogen™ Supplier No. PA551739

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MIGHIFEMND DDPHKEEEIR KYSAIYGRFD SKRKDGKHLT LHELTVNEAA AQLCVKDNAL LTRRDELFAL ARQISREVTY KYTYRTTKSK CGERDELSPK RIKVE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

Acts as a transcriptional repressor for zinc finger transcription factors EGR1 and EGR2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAB1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NAB1
Gene Accession No. Q13506
Gene Alias EGR1 binding protein 1; EGR-1-binding protein 1; Nab1; NGF1A-binding protein; Ngfi-A binding protein 1; NGFI-A binding protein 1 (EGR1 binding protein 1); NGFI-A-binding protein 1; Transcriptional regulatory protein p54
Gene Symbols NAB1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NAB1. Recombinant protein control fragment (Product #RP-102161).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4664
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.