missing translation for 'onlineSavingsMsg'
Learn More

Naftifine hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 15570584 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15570584 5 g 5g
15560584 1 g 1g
15580584 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code. 15570584 Supplier Thermo Scientific Acros Supplier No. 458660050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65473-14-5
Molecular Formula C21H21N·ClH
Molecular Weight (g/mol) 323.86
InChI Key OLUNPKFOFGZHRT-YGCVIUNWSA-N
Synonym naftifine hydrochloride, naftifine hcl, naftin, exoderil, naftifungin, n-trans-cinnamyl-n-methyl-1-naphthylmethyl amine hydrochloride, e-n-cinnamyl-n-methyl-1-naphthalenemethylamine hydrochloride, naftin tn, e-n-methyl-n-1-naphthylmethyl-3-phenyl-2-propen-1-amine-hydrochloride
PubChem CID 5281098
ChEBI CHEBI:7452
IUPAC Name (E)-N-methyl-N-(naphthalen-1-ylmethyl)-3-phenylprop-2-en-1-amine;hydrochloride
SMILES CN(CC=CC1=CC=CC=C1)CC2=CC=CC3=CC=CC=C32.Cl

Specifications

Melting Point 175.0°C to 179.0°C
Infrared Spectrum Authentic
Quantity 5 g
Solubility Information Solubility in water: 0.0068g/L ( 25°C ). Other solubilities: in ethanol 0.034g/L ( 25°C )
Formula Weight 323.86
Percent Purity 99%
Chemical Name or Material Naftifine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.