missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NAIP Recombinant Rabbit Monoclonal Antibody (6B9M10)

Product Code. 17729323
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17729323 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17729323 Supplier Invitrogen™ Supplier No. MA542731
  • ONLINE EXCLUSIVE PRICE
    528.00€
    395.65€ / 100µL
    Save 132.35€
    25% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: THP-1. Immunogen sequence: MATQQKASDE RISQFDHNLL PELSALLGLD AVQLAKELEE EEQKERAKMQ KGYNSQMRSE AKRLKTFVTY EPYSSWIPQE MAAAGFYFTG VKSGIQCFCC.

Neuronal apoptosis inhibitor protein (NAIP) was the first human inhibitor of apoptosis protein (IAP) identified and was discovered by its association with the neurodegenerative disorder spinal muscular atrophy. Members of the IAP family contain one to three copies of an approximately 70 amino acid motif termed baculovirus IAP repeat (BIR); these BIRs promote protein-protein interactions with various caspases such as caspase-3, -7, and -9 as well as members of the TRAF family of signal molecules (reviewed in 3). Unlike other IAPs however, NAIP requires ATP to bind caspase-9 and is not inhibited by the IAP-inhibiting molecule Smac/DIABLO, suggesting that NAIP is unique among the IAPs in its regulation of its activity. Finally, although only one human NAIP protein has been identified, other shorter NAIP mRNA transcripts have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAIP
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 6B9M10
Concentration 1.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NAIP
Gene Accession No. Q13075, Q9QWK5
Gene Alias AV364616; baculoviral IAP repeat-containing 1a; baculoviral IAP repeat-containing protein 1; Baculoviral IAP repeat-containing protein 1a; BIRC1; Birc1a; D13Lsd1; FLJ18088; FLJ42520; FLJ58811; Naip; Naip1; Naip-rs1; Neuronal apoptosis inhibitory protein; neuronal apoptosis inhibitory protein 1; NLR family apoptosis inhibitory protein; NLR family, apoptosis inhibitory protein; NLR family, apoptosis inhibitory protein 1; NLRB1; nucleotide-binding oligomerization domain, leucine rich repeat and BIR domain containing 1; psi neuronal apoptosis inhibitory protein; psiNAIP
Gene Symbols NAIP, Naip1
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17940, 4671
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.