missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NAM8 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17929212
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17929212 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17929212 Supplier Invitrogen™ Supplier No. PA5144487

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NAM8 Polyclonal Antibody for Western Blot, ELISA

Component of the U1 small nuclear ribonucleoprotein complex (U1 snRNP) involved in the initiation of meiotic recombination (PubMed:1603056, PubMed:7625279). Involved in the formation of DSBs at recombination hot-spots through meiosis-specific splicing of REC107 pre-mRNA (PubMed:7625279, PubMed:9112441, PubMed:21788335). Collaborates with MER1 to promote splicing of essential meiotic mRNAs REC10, AMA1, MER3, HFM1, SPO22 and PCH2 (PubMed:10983980, PubMed:21208980, PubMed:21788335). NAM8 interacts with the pre-mRNA downstream of the 5' splice site, in a region of non-conserved sequence and is required for efficient splicing of uncapped RNA precursor (PubMed:10072385).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAM8
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene NAM8
Gene Accession No. Q00539
Gene Alias MRE2; MUD15
Gene Symbols NAM8
Host Species Rabbit
Immunogen Recombinant Saccharomyces cerevisiae (strain ATCC 204508/S288c) (Baker's yeast) NAM8 protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 856486
Target Species Yeast
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.