missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene D8 1000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30308827
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30308827 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30308827 Supplier Dr. Ehrenstorfer Supplier No. DREGA09011117DI
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Naphthalene D8
CAS 1146-65-2
Solvent or Matrix Dichloromethane
Molecular Formula C10 D8
Quantity 1 mL
Synonym Naphthalene-1,2,3,4,5,6,7,8-d8, Naphthalene-d8 (6CI,8CI,9CI), Naphthalene (C10D8), Octadeuterionaphthalene, Octadeuteronaphthalene, Perdeuterated naphthalene, Perdeuterionaphthalene, Perdeuteronaphthalene, Naphthalene D8
InChI Formula InChI=1S/C10H8/c1-2-6-10-8-4-3-7-9(10)5-1/h1-8H/i1D,2D,3D,4D,5D,6D,7D,8D
SMILES [2H]c1c([2H])c([2H])c2c([2H])c([2H])c([2H])c([2H])c2c1[2H]
IUPAC Name 1,2,3,4,5,6,7,8-octadeuterionaphthalene
Molecular Weight (g/mol) 136.22
Formula Weight 136.1128 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.