missing translation for 'onlineSavingsMsg'
Learn More

NCAM-1/CD56 Antibody (rNCAM1/8655), Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18590548 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18590548 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18590548 Supplier Novus Biologicals Supplier No. NBP320806AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NCAM-1/CD56 Monoclonal antibody specifically detects NCAM-1/CD56 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCAM-1/CD56
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rNCAM1/8655
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias CD56, CD56/ NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM
Host Species Mouse
Immunogen Recombinant human NCAM-1/CD56 protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Membrane Vesicle Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4684
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.