missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAPH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11515-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the barr gene family and a regulatory subunit of the condensin complex. This complex is required for the conversion of interphase chromatin into condensed chromosomes. The protein encoded by this gene is associated with mitotic chromosomes, except during the early phase of chromosome condensation. During interphase, the protein has a distinct punctate nucleolar localization.Specifications
| NCAPH | |
| Polyclonal | |
| Unconjugated | |
| NCAPH | |
| Barren homolog protein 1, BRRN, BRRN1, CAP H, CAPH, Condensin complex subunit 2, HCAP H, KIAA0074, NCAPH, XCAP H homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 215387, 23397, 680089 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q15003, Q8C156 | |
| Ncaph | |
| NCAPH Fusion Protein Ag2076 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction