missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAPH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | NCAPH |
|---|---|
| Concentration | 0.15 mg/mL |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16818653
|
Proteintech
11515-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16808653
|
Proteintech
11515-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the barr gene family and a regulatory subunit of the condensin complex. This complex is required for the conversion of interphase chromatin into condensed chromosomes. The protein encoded by this gene is associated with mitotic chromosomes, except during the early phase of chromosome condensation. During interphase, the protein has a distinct punctate nucleolar localization.Specifications
| NCAPH | |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Rat, Human | |
| Q15003, Q8C156 | |
| 215387, 23397, 680089 | |
| NCAPH Fusion Protein Ag2076 | |
| Primary | |
| -20°C | |
| NCAPH |
| 0.15 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Barren homolog protein 1, BRRN, BRRN1, CAP H, CAPH, Condensin complex subunit 2, HCAP H, KIAA0074, NCAPH, XCAP H homolog | |
| Ncaph | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title