missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NCAPH2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15919644
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15919644 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15919644 Supplier Invitrogen™ Supplier No. PA566964

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: AAKLQDFHQWY LAAYADHADS RRLRRKGPSF ADMEVLYWTH VKEQLETLRK LQRREVAEQW LRPAEEDHLE DSLEDLGAAD DFLEPEEYME PEG Highest antigen sequence identity to the following orthologs - mouse 70%, rat 69%.

Structural maintenance of chromosomes (SMC) and non-SMC condensin proteins associate into complexes that have been implicated in the process of chromosome condensation. A crucial prerequisite for accurate segregation of replicated sister chromatids is the condensation of the chromosomes into a manageable form prior to metaphase. The condensin I complex consists of two SMC subunits, SMC2 and SMC4, and three non-SMC subunits, CAP-H, CAP-G, and CAP-D2. An alternative complex, the condensin II complex, contains alternate non-SMC subunits, CAP-G2, CAP-H2, and CAP-D3. CAP-H2 is also known as Non-SMC condensin II complex, subunit H2 (NCAPH2) or kleisin beta isoform 2. The three non-SMC subunits in the condensing complexes form a regulatory subcomplex that is required to activate the SMC ATPases and to promote mitosis-specific chromatin binding of the holocomplex. The precise individual functions of each non-SMC protein in activation remain to be determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NCAPH2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NCAPH2
Gene Accession No. Q6IBW4
Gene Alias CAPH2; CAP-H2 subunit of the condensin II complex; Chromosome-associated protein H2; condensin-2 complex subunit H2; CTA-384D8.36; D15Ertd785e; hCAP-H2; kleisin beta; Kleisin-beta; Ncaph2; Non-SMC condensin II complex subunit H2; non-SMC condensin II complex, subunit H2
Gene Symbols NCAPH2
Host Species Rabbit
Immunogen Recombinant Human NCAPH2. Recombinant protein control fragment (Product #RP-105440).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29781
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.