missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDP Polyclonal Antibody
GREENER_CHOICE

Product Code. 16578851
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16578851 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16578851 Supplier Invitrogen™ Supplier No. PA596013

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: KTDSSFIMDS DPRRCMRHHY VDSISHPLYK CSSKMVLLAR CEGHCSQASR SEPLVSFSTV LKQPFRSSCH CCRPQTSKLK ALRLRCSGGM RLTATYRYIL SCHCEECNS; Positive Samples: Mouse liver, Mouse heart, Mouse eye.

ALDH4A1 belongs to the aldehyde dehydrogenase family of proteins. This enzyme is a mitochondrial matrix NAD-dependent dehydrogenase which catalyzes the second step of the proline degradation pathway, converting pyrroline-5-carboxylate to glutamate. Deficiency of this enzyme is associated with type II hyperprolinemia, an autosomal recessive disorder characterized by accumulation of delta-1-pyrroline-5-carboxylate (P5C) and proline.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NDP
Gene Accession No. P48744
Gene Alias EVR2; FEVR; ND; NDP; NDP, norrin cystine knot growth factor; Ndph; Norrie disease (pseudoglioma); Norrie disease (pseudoglioma) (human); Norrie disease homolog; norrie disease protein; Norrie disease protein homolog; norrin; norrin cystine knot growth factor NDP; RGD1563968; similar to norrin; X-linked exudative vitreoretinopathy 2 protein
Gene Symbols NDP
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 25-133 of human NDP (NP_0002571).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17986
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.