Learn More
Negative Control for Mouse IgG1 Ab-1, Epredia™
Provides accurate, reproducible results with the Epredia™ Negative Control for Mouse IgG1 Ab-1, Mouse Monoclonal Antibody.
Marca: Epredia NC-748-P1ABX
Descrizione
This mouse monoclonal IgG1 was produced against a synthetic hapten, which is normally not present in humans or animals. Therefore, this mouse IgG1 does not react with any antigen of human or animal cells. This monoclonal IgG1 was purified from mouse ascites produced under the same experimental conditions as the other hybridoma cell lines of NeoMarkers. In order to serve as a true negative control, this IgG1 should be used under the identical conditions and at the same concentration as NeoMarkers' other purified mouse monoclonal IgG1 antibodies.Host Species: Mouse
Clone: NCG01
Isotype: IgG1/κ
Species Reactivity: All species.
Recommended for:
Negative control for:
- Immunohistochemistry (Frozen and formalin/paraffin)
- Western Blotting
- Immunofluorescence
- Immunoprecipitation
- Flow Cytometry
- ELISA
- Gel Supershift
Note:
USA: RUO; Int'l: RUO
Specifica
| Negative Control for Mouse IgG1 Ab-1 | |
| Monoclonal | |
| 1mg/mL | |
| Mouse | |
| RUO | |
| Primary | |
| Concentrated |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| NCG01 | |
| Unconjugated | |
| 100 μL | |
| Negative Controls | |
| All species | |
| IgG2a κ |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.