missing translation for 'onlineSavingsMsg'
Learn More

Negative Control for Mouse IgG1 Ab-1, Epredia™

Provides accurate, reproducible results with the Epredia™ Negative Control for Mouse IgG1 Ab-1, Mouse Monoclonal Antibody.

129.54€ - 443.00€

Specifications

Antigen Negative Control for Mouse IgG1 Ab-1
Clone NCG01
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 6
Product Code Brand Quantity Concentration Price Quantity & Availability  
Product Code Brand Quantity Concentration Price Quantity & Availability  
12622687
View Documents
Epredia
NC-748-P0
100 μL 200μg/mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12642687
View Documents
Epredia
NC-748-P1ABX
100 μL 1mg/mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12652687
View Documents
Epredia
NC-748-PABX
200 μL 1mg/mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12632687
View Documents
Epredia
NC-748-P1
500 μL 200μg/mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12612687
View Documents
Epredia
NC-748-P
1 mL 200μg/mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
12672687
View Documents
Epredia
NC-748-R7
7 mL Prediluted This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
Description

Description

This mouse monoclonal IgG1 was produced against a synthetic hapten, which is normally not present in humans or animals. Therefore, this mouse IgG1 does not react with any antigen of human or animal cells. This monoclonal IgG1 was purified from mouse ascites produced under the same experimental conditions as the other hybridoma cell lines of NeoMarkers. In order to serve as a true negative control, this IgG1 should be used under the identical conditions and at the same concentration as NeoMarkers' other purified mouse monoclonal IgG1 antibodies.

Host Species: Mouse

Clone: NCG01

Isotype: IgG1/κ

Species Reactivity: All species.

Recommended for:

Negative control for:

  • Immunohistochemistry (Frozen and formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • ELISA
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Negative Control for Mouse IgG1 Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RUO
All species
Primary
NCG01
Monoclonal
Mouse
Negative Controls
IgG2a κ
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.