missing translation for 'onlineSavingsMsg'
Learn More

Negative Control for Mouse IgG1 Ab-1, Epredia™

Provides accurate, reproducible results with the Epredia™ Negative Control for Mouse IgG1 Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia NC-748-R7

 View more versions of this product

Product Code. 12672687

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »

This item is not returnable. View return policy

Description

Description

This mouse monoclonal IgG1 was produced against a synthetic hapten, which is normally not present in humans or animals. Therefore, this mouse IgG1 does not react with any antigen of human or animal cells. This monoclonal IgG1 was purified from mouse ascites produced under the same experimental conditions as the other hybridoma cell lines of NeoMarkers. In order to serve as a true negative control, this IgG1 should be used under the identical conditions and at the same concentration as NeoMarkers' other purified mouse monoclonal IgG1 antibodies.

Host Species: Mouse

Clone: NCG01

Isotype: IgG1/κ

Species Reactivity: All species.

Recommended for:

Negative control for:

  • Immunohistochemistry (Frozen and formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Flow Cytometry
  • ELISA
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Negative Control for Mouse IgG1 Ab-1
Monoclonal
Prediluted
Mouse
RUO
Primary
Prediluted
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
NCG01
Unconjugated
7 mL
Negative Controls
All species
IgG2a κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.