Learn More
Neostigmine bromide, Thermo Scientific™
A reversible acetylcholinesterase inhibitor
Brand: Thermo Scientific Alfa Aesar J62300.03
| Quantity | 1g |
|---|
View more versions of this product
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 114-80-7 | |
| 303.20 | |
| LULNWZDBKTWDGK-UHFFFAOYSA-M | |
| 8246 | |
| 3-[(dimethylcarbamoyl)oxy]-N,N,N-trimethylanilinium bromide |
| C12H19BrN2O2 | |
| MFCD00011795 | |
| neostigmine bromide, neo-proserin, eustigmin bromide, neoserine bromide, proserine bromide, stigmanol bromide, synstigmin bromide, leostigmine bromide, kirkstigmine bromide, philostigmin bromide | |
| CHEBI:179557 | |
| [Br-].CN(C)C(=O)OC1=CC=CC(=C1)[N+](C)(C)C |
Specifications
| 114-80-7 | |
| MFCD00011795 | |
| UN2811 | |
| neostigmine bromide, neo-proserin, eustigmin bromide, neoserine bromide, proserine bromide, stigmanol bromide, synstigmin bromide, leostigmine bromide, kirkstigmine bromide, philostigmin bromide | |
| [Br-].CN(C)C(=O)OC1=CC=CC(=C1)[N+](C)(C)C | |
| 303.20 | |
| CHEBI:179557 | |
| Neostigmine bromide |
| C12H19BrN2O2 | |
| 1g | |
| 14,6464 | |
| LULNWZDBKTWDGK-UHFFFAOYSA-M | |
| 3-[(dimethylcarbamoyl)oxy]-N,N,N-trimethylanilinium bromide | |
| 8246 | |
| 303.2 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.