Learn More
Neostigmine bromide, Thermo Scientific™
A reversible acetylcholinesterase inhibitor
44.30€ - 71.90€
Chemical Identifiers
| CAS | 114-80-7 |
|---|---|
| Molecular Formula | C12H19BrN2O2 |
| Molecular Weight (g/mol) | 303.20 |
| MDL Number | MFCD00011795 |
| InChI Key | LULNWZDBKTWDGK-UHFFFAOYSA-M |
| Synonym | neostigmine bromide, neo-proserin, eustigmin bromide, neoserine bromide, proserine bromide, stigmanol bromide, synstigmin bromide, leostigmine bromide, kirkstigmine bromide, philostigmin bromide |
| PubChem CID | 8246 |
| ChEBI | CHEBI:179557 |
| IUPAC Name | 3-[(dimethylcarbamoyl)oxy]-N,N,N-trimethylanilinium bromide |
| SMILES | [Br-].CN(C)C(=O)OC1=CC=CC(=C1)[N+](C)(C)C |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15414709
|
Thermo Scientific Alfa Aesar
J62300.MD |
250mg |
44.30€
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15404709
|
Thermo Scientific Alfa Aesar
J62300.03 |
1g |
71.90€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 114-80-7 | |
| 303.20 | |
| LULNWZDBKTWDGK-UHFFFAOYSA-M | |
| 8246 | |
| 3-[(dimethylcarbamoyl)oxy]-N,N,N-trimethylanilinium bromide |
| C12H19BrN2O2 | |
| MFCD00011795 | |
| neostigmine bromide, neo-proserin, eustigmin bromide, neoserine bromide, proserine bromide, stigmanol bromide, synstigmin bromide, leostigmine bromide, kirkstigmine bromide, philostigmin bromide | |
| CHEBI:179557 | |
| [Br-].CN(C)C(=O)OC1=CC=CC(=C1)[N+](C)(C)C |
Specifications
| 114-80-7 | |
| MFCD00011795 | |
| 14,6464 | |
| LULNWZDBKTWDGK-UHFFFAOYSA-M | |
| 3-[(dimethylcarbamoyl)oxy]-N,N,N-trimethylanilinium bromide | |
| 8246 | |
| 303.2 |
| C12H19BrN2O2 | |
| UN2811 | |
| neostigmine bromide, neo-proserin, eustigmin bromide, neoserine bromide, proserine bromide, stigmanol bromide, synstigmin bromide, leostigmine bromide, kirkstigmine bromide, philostigmin bromide | |
| [Br-].CN(C)C(=O)OC1=CC=CC(=C1)[N+](C)(C)C | |
| 303.20 | |
| CHEBI:179557 | |
| Neostigmine bromide |
Safety and Handling
GHS H Statement
H300-H310-H330-H334-H315-H319-H317-H335
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P501c
H301-H315-H317-H319-H334-H335
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 204-054-8
missing translation for 'rtecsNumber' : BR3150000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.