missing translation for 'onlineSavingsMsg'
Learn More

Nesfatin-1/Nucleobindin-2 Antibody, Novus Biologicals™

Product Code. 18746133 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18746133 0.1 mL 0.1mL
18421241 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18746133 Supplier Novus Biologicals Supplier No. NBP187383

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 4 publications

Nesfatin-1/Nucleobindin-2 Polyclonal specifically detects Nesfatin-1/Nucleobindin-2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY

Specifications

Applications Western Blot, Immunohistochemistry, Immunofluorescence
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Knockdown Validated
Purification Method Affinity Purified
Quantity 0.1 mL

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.