Learn More
NEU2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24523-1-AP-20UL
 View more versions of this product
Product Code. 16882995
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
The NEU2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human NEU2. This antibody recognizes human, mouse, rat antigen. The NEU2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
NEU2
Polyclonal
Unconjugated
20 μL
4759
IgG
Western Blot, ELISA
267 μg/mL
Rabbit
Primary
Human, Rat, Mouse