Learn More
NEU2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | NEU2 |
|---|---|
| Concentration | 267 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
The NEU2 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human NEU2. This antibody recognizes human, mouse, rat antigen. The NEU2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| NEU2 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human, Rat, Mouse | |
| IgG |
| 267 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 4759 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.