missing translation for 'onlineSavingsMsg'
Learn More

Neurogenin-3 Antibody (PCRP-NEUROG3-1E10), mFluor Violet 610 SE, Novus Biologicals™

Product Code. 18562025 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18562025 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18562025 Supplier Novus Biologicals Supplier No. NBP314029MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Neurogenin-3 Monoclonal antibody specifically detects Neurogenin-3 in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen Neurogenin-3
Applications Flow Cytometry, ELISA, Immunoprecipitation, Protein Array
Classification Monoclonal
Clone PCRP-NEUROG3-1E10
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, ELISA, Immunoprecipitation, Protein Array
Formulation 50mM Sodium Borate
Gene Alias Atoh5, BHLHA7, bHLHa7NGN-3, Class A basic helix-loop-helix protein 7, Math4B, neurogenin 3, neurogenin-3, NGN3, Protein atonal homolog 5
Host Species Mouse
Immunogen Recombinant human full-lengthNeurogenin-3protein (Uniprot: Q9Y4Z2)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 50674
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.