missing translation for 'onlineSavingsMsg'
Learn More

Neuropilin 1 Mouse anti-Human, Porcine, Clone: 2H3F6, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60067-1-IG-20UL

 View more versions of this product

Product Code. 16817035

  • 145.00€ / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Neuropilin 1 is a membrane-bound coreceptor to a tyrosine kinase receptor for both vascular endothelial growth factor and semaphorin family members. Neuropilin 1 plays versatile roles in angiogenesis, axon guidance, cell survival, migration and invasion.
Specifications

Specifications

Neuropilin 1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O14786
NRP1
Neuropilin 1 Fusion Protein Ag0931
20 μL
Primary
Pig, Human
Antibody
IgG1
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
2H3F6
Unconjugated
NRP1
BDCA4, CD304, neuropilin 1, NRP, NRP1, VEGF165R
Mouse
Caprylic Acid Precipitation
RUO
100286859, 8829
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.