missing translation for 'onlineSavingsMsg'
Learn More

Neuropilin 1 Mouse anti-Human, Porcine, Clone: 2H3F6, Proteintech

Mouse Monoclonal Antibody

145.00€ - 410.00€

Specifications

Antigen Neuropilin 1
Clone 2H3F6
Concentration 1 mg/mL
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16817035
View Documents
Proteintech
60067-1-IG-20UL
20 μL
145.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16807035
View Documents
Proteintech
60067-1-IG-150UL
150 μL
410.00€
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Neuropilin 1 is a membrane-bound coreceptor to a tyrosine kinase receptor for both vascular endothelial growth factor and semaphorin family members. Neuropilin 1 plays versatile roles in angiogenesis, axon guidance, cell survival, migration and invasion.
Specifications

Specifications

Neuropilin 1
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
BDCA4, CD304, neuropilin 1, NRP, NRP1, VEGF165R
NRP1
IgG1
Caprylic Acid Precipitation
Antibody
2H3F6
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Mouse
Pig, Human
O14786
100286859, 8829
Neuropilin 1 Fusion Protein Ag0931
Primary
-20°C
NRP1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.