missing translation for 'onlineSavingsMsg'
Learn More

NFATC1/NFAT2 Antibody (7A6), Novus Biologicals™

Product Code. 18789263 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18789263 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18789263 Supplier Novus Biologicals Supplier No. NB300620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

NFATC1/NFAT2 Monoclonal specifically detects NFATC1/NFAT2 in Human, Mouse, Rat, Hamster, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Gel Super Shift Assays.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFATC1/NFAT2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 7A6
Concentration 9.25mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:2000, Simple Western, Immunohistochemistry 1:20 - 1:200, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:20 - 1:200, Gel Super Shift Assays 1:1 - 1:100, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Gene Accession No. O95644
Gene Alias MGC138448, NFAT transcription complex cytosolic component, NFAT2cytoplasmic 1, NFATC, NF-ATC, NFATc1, NF-ATc1, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1
Gene Symbols NFATC1
Host Species Mouse
Immunogen Bacterially expressed glutathione S-transferase (GST) fusion protein containing NF-ATc1 residues 1-654.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 4772
Test Specificity Detects cytosolic nuclear factor of activated T-cells (NFAT 2) from transfected human, mouse, and monkey cells. This does not cross react with NFATp.
Target Species Human, Mouse, Rat, Hamster, Primate, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.