missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinic Acetylcholine R alpha 1/CHRNA1 Antibody (mAb 192), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Nicotinic Acetylcholine R alpha 1/CHRNA1 Monoclonal antibody specifically detects Nicotinic Acetylcholine R alpha 1/CHRNA1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Neutralization
Specifications
Specifications
| Applications | Western Blot, ELISA |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Block/Neutralize |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Gene ID (Entrez) | 1134 |
| Isotype | IgG2b κ |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?