Learn More
Nitric acid, 65-70%, 99.999% (metals basis), Thermo Scientific Chemicals
Beskrivning
In the manufacture of fertilizers and explosives, in organic synthesis, in metallurgy, in photoengraving, in etching steel, in rubber chemicals, and in reprocessing nuclear fuelsMain commercial usage is in the production of fertilizers and reagent for nitration. It implies in the applications for the production of explosives, nylon precursors, and specialty organic compounds. Nitric acid is indirectly used as an oxidizer in liquid-fueled rockets for determining trace level metals in solutions. It is used as a chemical doping agent for organic semiconductors, and in purification processes for raw carbon nanotubes, artificially age pine and maple.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifikationer
Specifikationer
| Color | Colorless to Yellow |
| Boiling Point | 121°C |
| Density | 1.41 |
| Odor | Pungent |
| Quantity | 100 mL |
| Physical Form | Liquid |
| UN Number | UN2031 |
| Merck Index | 14,6577 |
| Solubility Information | Very soluble in water and ether; Decomposes in alcohol;Miscible with water and ether. |
| Formula Weight | 63.01 |
| Visa mer |
RUO – Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.