missing translation for 'onlineSavingsMsg'
Learn More

Nitrofurantoin 13C3, Dr. Ehrenstorfer

Product Code. 30312147
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312147 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312147 Supplier Dr. Ehrenstorfer Supplier No. DREC15570910
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nitrofurantoin 13C3
CAS 1217226-46-4
Molecular Formula 13C3 C5 H6 N4 O5
Quantity 5 mg
InChI Formula InChI=1S/C8H6N4O5/c13-6-4-11(8(14)10-6)9-3-5-1-2-7(17-5)12(15)16/h1-3H,4H2,(H,10,13,14)/b9-3+/i4+1,6+1,8+1
SMILES [O-][N+](=O)c1oc(\C=N\N2[13CH2][13C](=O)N[13C]2=O)cc1
IUPAC Name 1-[(E)-(5-nitrofuran-2-yl)methylideneamino]-(2,4,5-^{13}C_{3})1,3-diazolidine-2,4-dione
Molecular Weight (g/mol) 241.14
Formula Weight 241.0439 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.