missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NK1.1 (CD161) Mouse, anti-Mouse, Biotin, Clone: PK136, STEMCELL Technologies
Mouse Monoclonal Antibody
POA - 498.15€
Specifications
| Antigen | NK1.1 (CD161) |
|---|---|
| Clone | PK136 |
| Applications | CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay |
| Classification | Monoclonal |
| Conjugate | Biotin |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17197621
|
Stemcell Technologies
100-0462 |
25 μg | |||||||
17107631
|
Stemcell Technologies
100-0463 |
100 μg |
243.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17189011
|
Stemcell Technologies
60103BT |
500 μg |
498.15€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- The PK136 antibody reacts with murine NK1.1 (CD161), an ∼80 kDa homodimeric type 2 transmembrane glycoprotein and C-type lectin receptor expressed on NK cells, NK-T cells, and rare subsets of T cells and monocytes in select strains of mice
- NK1.1 is encoded by the Klrb1b and Klrb1c genes, which specify CD161b and CD161c polypeptides, respectively
- CD161b is expressed only by Swiss NIH and SJL mice, and CD161c by certain strains such as C57BL, FVB/N, and NZB (but not A, AKR, BALB/c, CBA/J, C3H, C57BR, C58, DBA/1, DBA/2, NOD, SJL, or 129)
- NK1.1 has functional roles in modulating several processes, including the activation and proliferation of NK cells, induction of interferon-γ production, and release of cytotoxic granules
- Its expression on the cell surface is specifically upregulated by IL-12
- For detection of NK cells in NK1.1- strains, Anti-Mouse CD49b Antibody, Clone DX5 (Catalog #60020) is recommended
- DX5 recognizes CD49b (integrin α2), another commonly used NK cell marker
Specifications
| NK1.1 (CD161) | |
| CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Functional Assay | |
| Biotin | |
| Mouse |
| PK136 | |
| Monoclonal | |
| Mouse | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title