missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2.2 Mouse, Clone: NX2/1523, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 4821-MSM3-P0
This item is not returnable.
View return policy
Specifications
| NKX2.2 | |
| Nuclear | |
| NX2/1523 | |
| Unconjugated | |
| XP_006723629 | |
| Mouse | |
| 40-50kDa | |
| Protein A/G Purified | |
| RUO | |
| 4821 | |
| IgG1 κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| NKX2-2 | |
| Human recombinant NKX2.2 protein fragment (aa1-119) | |
| Pancreas or Ewing s sarcoma | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction