missing translation for 'onlineSavingsMsg'
Learn More

NKX2.2 Mouse, Clone: NX2/1523, enQuireBio™

Mouse Monoclonal Antibody

199.00€ - 478.00€

Specifications

Antigen NKX2.2
Clone NX2/1523
Concentration 200μg/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15910588
View Documents
EnquireBio
4821-MSM3-P0
20 μg This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
N/A View alternative products
Save Up to 
»
15920588
View Documents
EnquireBio
4821-MSM3-P1
100 μg
ONLINE EXCLUSIVE PRICE
554.40€
478.00€ / 100µg
Save 76.40€
13% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NKX2.2
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% Azide
4821
Human recombinant NKX2.2 protein fragment (aa1-119)
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
40-50kDa
NX2/1523
Immunohistochemistry
Unconjugated
RUO
XP_006723629
NKX2-2
IgG1 κ
Protein A/G Purified
Nuclear
Pancreas or Ewing s sarcoma
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.