missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nkx2.4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15673975
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15673975 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15673975 Supplier Invitrogen™ Supplier No. PA550046

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total NKX2-4 protein.

NKX2-4(NK2 Homeobox4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includesequence-specific DNA binding and may have transcription factor function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx2.4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NKX2-4
Gene Accession No. Q9H2Z4
Gene Alias 1700001P03Rik; Homeobox protein NK-2 homolog D; homeobox protein Nkx-2.4; LOW QUALITY PROTEIN: homeobox protein Nkx-2.4; NK2 homeobox 1a; NK2 homeobox 4; NK2 transcription factor homolog D; NK2 transcription factor related, locus 4; NKX2.4; Nkx-2.4; NKX2-4; NKX2D; tinman
Gene Symbols NKX2-4
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human NKX24.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644524
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.