missing translation for 'onlineSavingsMsg'
Learn More

NLG919-d10, TRC

Product Code. 30477773
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30477773 10 mg 10mg
30478994 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30477773 Supplier Toronto Research Chemicals Supplier No. TRCN63000210MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material NLG919-d10
Molecular Formula C18H12D10N2O
Quantity 10 mg
Synonym α-Cyclohexyl-5H-imidazo[5,1-a]isoindole-5-ethanol-d10, 1-Cyclohexyl-2-(5H-imidazo[5,1-a]isoindol-5-yl)ethanol-d10
InChI Formula InChI=1S/C18H22N2O/c21-18(13-6-2-1-3-7-13)10-16-14-8-4-5-9-15(14)17-11-19-12-20(16)17/h4-5,8-9,11-13,16,18,21H,1-3,6-7,10H2/i1D2,2D2,3D2,6D2,7D2
SMILES OC(C1C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H])CC2N3C(C4=C2C=CC=C4)=CN=C3
Molecular Weight (g/mol) 292.44
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.