missing translation for 'onlineSavingsMsg'
Learn More

METTL7A Mouse anti-Human, Clone: OTI4B10, liquid, TrueMAB™

Product Code. 17582494
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17582494 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17582494 Supplier Origene Technologies Supplier No. TA809878
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Antibody

METTL7A is a methyltransferase-like protein and members of this family are SAM dependent methyltransferases.

Specifications

Antigen METTL7A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4B10
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene METTL7A
Gene Accession No. Q9H8H3
Gene Alias AAM-B
Gene Symbols METTL7A
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 34-104 of human METTL7A produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25840
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.