Learn More
NLRP3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19771-1-AP-20UL
Description
The NLRP3 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human NLRP3. This antibody recognizes human antigen. The NLRP3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| NLRP3 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 114548 | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP), ELISA | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human |