Learn More
NLRP3 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | NLRP3 |
|---|---|
| Concentration | 333 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP), ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16819254
|
Proteintech
19771-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16809254
|
Proteintech
19771-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NLRP3 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human NLRP3. This antibody recognizes human antigen. The NLRP3 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| NLRP3 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Co-immunoprecipitation (Co-IP), ELISA | |
| Unconjugated | |
| Human | |
| IgG |
| 333 μg/mL | |
| Polyclonal | |
| Rabbit | |
| 114548 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.