missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NOM1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15702041 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15702041 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15702041 Supplier Invitrogen™ Supplier No. PA554102

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ITDKSLCGSG EKYIPPHVRQ AEETVDFKKK EELERLKKHV KGLLNRLSEP NMASISGQLE ELYMAHSRKD MNDTLTSALM GACVTASAMP SRLMMEHVLL VSILHHTVGI EVGAHFLEAV VRKFDAIYKY GSE Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 77%.

Proteins that contain MIF4G (middle of eIF4G (MIM 600495)) and/or MA3 domains, such as NOM1, function in protein translation. These domains include binding sites for members of the EIF4A family of ATP-dependent DEAD box RNA helicases (see EIF4A1; MIM 602641) (Simmons et al., 2005 [PubMed 15715967]).[supplied by OMIM, Mar 2008].
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen NOM1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NOM1
Gene Accession No. Q5C9Z4
Gene Alias C7orf3; D5Kng1; Gm1040; Nom1; Nucleolar MIF4G domain-containing protein 1; nucleolar protein with MIF4G domain 1; PPP1R113; protein phosphatase 1, regulatory subunit 113; SGD1; SGD1 homolog
Gene Symbols NOM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NOM1. Recombinant protein control fragment (Product #RP-92761).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64434
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.