Learn More
NOXA2/p67phox Mouse, anti-Human, CoraLite Plus 488, Clone: 1D11A3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-67594-100UL
 View more versions of this product
Product Code. 18014805
  • 450.00€ / 100µL
Description
The NOXA2/p67phox antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NOXA2/p67phox. This antibody recognizes human antigen. The NOXA2/p67phox antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
NOXA2/p67phox
1D11A3
CoraLite Plus 488
Mouse
Primary
Human
155 kDa
IgG1
Immunofluorescence
1000 μg/mL
FLJ93058, NADPH oxidase activator 2, NCF 2, NCF2, Neutrophil cytosol factor 2, neutrophil cytosolic factor 2, NOXA2, P67 PHOX, P67PHOX
100 μL
4688
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.