Learn More
NOXA2/p67phox Mouse, anti-Human, CoraLite Plus 488, Clone: 1D11A3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67594-100UL
Description
The NOXA2/p67phox antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NOXA2/p67phox. This antibody recognizes human antigen. The NOXA2/p67phox antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| NOXA2/p67phox | |
| 1D11A3 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| FLJ93058, NADPH oxidase activator 2, NCF 2, NCF2, Neutrophil cytosol factor 2, neutrophil cytosolic factor 2, NOXA2, P67 PHOX, P67PHOX | |
| 100 μL | |
| 4688 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |