Learn More
NOXA2/p67phox Mouse, anti-Human, CoraLite Plus 488, Clone: 1D11A3, Proteintech
Mouse Monoclonal Antibody
145.00€ - 450.00€
Specifications
| Antigen | NOXA2/p67phox |
|---|---|
| Clone | 1D11A3 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18014805
|
Proteintech
CL488-67594-100UL |
100 μL |
450.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18028965
|
Proteintech
CL488-67594-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NOXA2/p67phox antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NOXA2/p67phox. This antibody recognizes human antigen. The NOXA2/p67phox antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| NOXA2/p67phox | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Mouse | |
| 4688 | |
| IgG1 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 1D11A3 | |
| Immunofluorescence | |
| Liquid | |
| Human | |
| FLJ93058, NADPH oxidase activator 2, NCF 2, NCF2, Neutrophil cytosol factor 2, neutrophil cytosolic factor 2, NOXA2, P67 PHOX, P67PHOX | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.