missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NPRL3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15720801 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15720801 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15720801 Supplier Invitrogen™ Supplier No. PA552849

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GDSRFSDVIL ATILATKSEM CGQKFELKID NVRFVGHPTL LQHALGQISK TDPSPKREAP TMILFNVVFA LRANADPSVI NCLHNLSRRI ATVLQHEERR CQYLTREAKL ILALQDEVSA MADGNEGPQS PFHHILPKCK Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.

The function of the encoded protein is not known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NPRL3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NPRL3
Gene Accession No. Q12980
Gene Alias -14 gene protein; Aag; alpha-globin regulatory element-containing gene protein; alphaRE-14; C16orf35; CGTHBA; conserved gene telomeric to alpha globin cluster; GATOR complex protein NPRL3; HS-26; HS40; HS-40; MARE; nitrogen permease regulator 3-like protein; NPR3; NPR3 like, GATOR1 complex subunit; NPR3-like; NPR3-like, GATOR1 complex subunit; NPRL3; Phg; Protein CGTHBA; Prox1; RMD11; RP23-177M10.6
Gene Symbols NPRL3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NPRL3. Recombinant protein control fragment (Product #RP-88735).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8131
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.