missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NPS 2390
GREENER_CHOICE

Product Code. 18015832
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18015832 50 mg 50mg
18065452 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18015832 Supplier Tocris Bioscience™ Supplier No. 4134/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Group I mGlu antagonist

Group I mGlu antagonist; displays noncompetitive antagonist activity at both mGlu1 and mGlu5 receptors. Thought to act on a site separate from the glutamate binding pocket.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 226878-01-9
Molecular Formula C19H21N3O

Specifications

Quantity 50 mg
Formula Weight 307.39
Purity >99%
Molecular Weight (g/mol) 307.397
Chemical Name or Material NPS 2390
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.