missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NuMA Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, HeLa, HepG2, HeLa, HeLa (100 ng/mL Nocodazole treatment for 24 hr). Predicted reactivity: Mouse (81%), Rat (83%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NuMA (nuclear mitotic apparatus) is a long coil-coiled protein that plays a role in the interphase and mitosis phases of a cell cycle. In interphase, it has been hypothesized to play a structural role in the nucleoskeleton that may be important in nuclear organization and functions. During mitosis, it is associated with spindle microtubule organization and chromosome positioning.
Specifications
Specifications
| Antigen | NuMA |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.54 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | NUMA1 |
| Gene Accession No. | Q14980 |
| Gene Alias | 6720401E04Rik; AA764025; AL022610; AU014979; centrophilin stabilizes mitotic spindle in mitotic cells; NMP22; NMP-22; Nuclear matrix protein-22; Nuclear mitotic apparatus protein; Nuclear mitotic apparatus protein 1; NUMA; NuMA protein; Numa1; SP-H antigen; structural nuclear protein |
| Gene Symbols | NUMA1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?