Learn More
DWK Life Sciences O-ring, made of nitrile, for safety standard ground sockets
Product Code. 15991551 Shop All DWK Life Sciences Products
Change view
Click to view available options
Joint Size:
NS 14/23
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
Unit Size:
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Joint Size Unit Size
15991551 NS 19/26 Pack of 10
15921561 NS 45/40 Pack of 10
15864608 NS 14/23 Pack of 10
15884608 NS 29/32 Pack of 10
15894608 NS 34/35 Pack of 10
15911561 NS 24/29 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15991551 Supplier DWK Life Sciences Supplier No. 286217002
  • ONLINE EXCLUSIVE PRICE
    8.70€
    7.22€ / Pack of 10
    Save 1.48€
    17% Off
View all promotions
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Component of the DURAN™ safety ground joint.
Nitrile O-rings for DURAN™ safety ground joints together with the screw cap secure the connection between core and sleeve.
  • In combination with the screw cap, the safety ground joints (core and sleeve) can be sealed tightly.
Specifications
For Use With (Application) To secure the security grindings; Sealing
Joint Size NS 19/26
Quantity 10/Pk.
Material Nitrile