missing translation for 'onlineSavingsMsg'
Learn More

O-Tolidine Dihydrochloride, TRC

Product Code. 30449161
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30449161 2.5 g 2.5g
30455649 500 mg 500mg
30454483 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30449161 Supplier Toronto Research Chemicals Supplier No. TRCT7338852.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-82-8
Molecular Formula C14H16N2 . 2HCl
Molecular Weight (g/mol) 285.2
IUPAC Name 4-(4-amino-3-methylphenyl)-2-methylaniline;dihydrochloride
SMILES Cl.Cl.Cc1cc(ccc1N)c2ccc(N)c(C)c2

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C14H16N2.2ClH/c1-9-7-11(3-5-13(9)15)12-4-6-14(16)10(2)8-12;;/h3-8H,15-16H2,1-2H3;2*1H
Formula Weight 284.0847
Chemical Name or Material O-Tolidine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.