missing translation for 'onlineSavingsMsg'
Learn More

OCIAD1 Mouse anti-Human, Mouse, Rat, Clone: 1C10C3, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66698-1-IG-150UL

 View more versions of this product

Product Code. 16898815

  • 410.00€ / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody detects OCIAD1 with an apparent molecular weight of ∽35 kDa as has been demonstrated by several researches.

The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab subfamilies, exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. At least eight members of the Rab subfamily have been identified, each of which is found at a particular stage of a membrane transport pathway.
Specifications

Specifications

OCIAD1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5XIG4, Q9CRD0, Q9NX40
Ociad1
OCIAD1 Fusion Protein Ag9977
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG2a
Western Blot
1C10C3
Unconjugated
OCIAD1
Asrij, FLJ20455, OCIA, OCIA domain containing 1, OCIAD1, TPA018
Mouse
Protein A
RUO
289590, 54940, 68095
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.