missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCIAD1 Mouse anti-Human, Mouse, Rat, Clone: 1C10C3, Proteintech
Mouse Monoclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | OCIAD1 |
|---|---|
| Clone | 1C10C3 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16808825
|
Proteintech
66698-1-IG-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16898815
|
Proteintech
66698-1-IG-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody detects OCIAD1 with an apparent molecular weight of ∽35 kDa as has been demonstrated by several researches.
The Ras-related superfamily of guanine nucleotide binding proteins, which includes the R-Ras, Rap, Ral/Rec and Rho/Rab subfamilies, exhibit 30-60% homology with Ras p21. Accumulating data suggests an important role for Rab proteins, either in endocytosis or in biosynthetic protein transport. At least eight members of the Rab subfamily have been identified, each of which is found at a particular stage of a membrane transport pathway.Specifications
| OCIAD1 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Asrij, FLJ20455, OCIA, OCIA domain containing 1, OCIAD1, TPA018 | |
| Ociad1 | |
| IgG2a | |
| Protein A | |
| Antibody |
| 1C10C3 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Mouse, Rat, Human | |
| Q5XIG4, Q9CRD0, Q9NX40 | |
| 289590, 54940, 68095 | |
| OCIAD1 Fusion Protein Ag9977 | |
| Primary | |
| -20°C | |
| OCIAD1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title